Learn More
Bochem™ 18/10 Stainless Steel Chemical Spoon
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
32 mm
40 mm
45 mm
48 mm
Diameter (Metric) Outer:
1 mm
2 mm
5 mm
Unit Size:
Each
Pack of 10
Description
- Chemical spoon
- With spatula handle
Specifications
Specifications
| Quantity | 10 Pk. |
| Product Type | Chemical Scoop |
| Length (Metric) Blade | 40 mm |
| Width (Metric) | 29 mm |
| Diameter (Metric) Outer | 5 mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 210 mm |