missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A Tyvek™ Zipper Equipment Cover
GREENER_CHOICE

1422A Tyvek™ Zipper Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.

€ 4315.00 - € 7915.00


Products 5
Product Code Brand Length (English) Width (English) Height (English) Price Quantity & Availability  
Product Code Brand Length (English) Width (English) Height (English) Price Quantity & Availability  
18604979
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-262650Z2
26 in. 26 in. 50 in.
€ 4360.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
17758277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-302456Z1
30 in. 24 in. 56 in.
€ 4490.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
18614979
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-322244Z2
32 in. 22 in. 44 in.
€ 4315.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
17708307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-502872Z2WP4
50 in. 28 in. 72 in.
€ 4900.00
Pack of 50
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
 
17758307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-545480Z2CP4
€ 7915.00
Pack of 50
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
 
Description

Description

  • Keeps Cleaned Equipment Clean and Protected from Contamination
  • Breathable
  • Form Fitting, Fast and Consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Keystone Cleanroom Products 1422A Tyvek™ Zipper Equipment Cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.