Learn More
Keystone Cleanroom Products 1422A Tyvek Autoclavable Bowl Cover, with Steam Indicator
Keystone Cleanroom Products 1422A Tyvek™ Autoclavable Bowl Covers prevent microbial and particulate contamination to critical parts
Brand: Keystone Cleanroom Products BWLT2A24SP250
This item is not returnable.
View return policy
Description
- Prevent microbial and particulate contamination to critical parts, while the fill line set-up activities are performed
- Manufactured and packaged in a controlled cleanroom environment
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Steam Sterilization Indicator - Autoclavable at 121°C to 125°C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
Specifications
Autoclavable | |
White | |
Steam Sterilization Indicator | |
250/Pk. | |
24 in. |
Certificate of Conformance (COC) | |
Steam Sterilization | |
Tyvek | |
121°C to 125°C | |
61 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Keystone Cleanroom Products 1422A Tyvek Autoclavable Bowl Cover, with Steam Indicator > 12 in. to 20 in.; 250/Pk.