Learn More
Helios Italquartz 1234 DIN Standard Crucibles, Transparent
1234 DIN standard crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 63P00506
 View more versions of this product
Product Code. 15848054
  • € 168.00 / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Description
  • Transparent
  • Conform to 1234 DIN Standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
Quartz
60 mm
1234 DIN standard crucible
Model CR 46
50 cc
38 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Helios Italquartz 1234 DIN Standard Crucibles, Transparent > 50 cc, 60 mm OD, 38 mm high