Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
Product Code. 10109560
Click to view available options
Capacity (Metric):
300 mL
1 L
1500 mL
2 L
Includes:
No side grip
With side grip
Height (Metric) Inner:
190mm
235mm
240mm
245mm
290mm
340mm
Diameter (Metric) Inner:
47 mm
77 mm
90 mm
100 mm
Unit Size:
Each
Product Code. 10109560
Brand: KGW Isotherm™ 10214
Only null left
Add to basket
Add to basket
For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.
  • Easy handling
  • Cooling of small samples
  • Fully silvered as standard
  • Flexible applications in laboratory
  • Available with or without side grip
Specifications
Color Blue
Diameter (Metric) Outer 115mm
For Use With (Application) Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask
Height (Metric) Inner 245mm
Height (Metric) 300mm
Includes No side grip
Capacity (Metric) 1500 mL
Diameter (Metric) Inner 90 mm
Material Borosilicate Glass