Learn More
Thermo Scientific™ 100% Graphite Ferrules
Product Code. p-3336780 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
For Varian, Perkin-Elmer
Description:
0.25 in. O.D.
Unit Size:
Pack of 10
Product Code. 10659665
Brand: Thermo Scientific™ 29033405
Only null left
Add to basket
Add to basket
Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections
  • Easy-to-use
  • Form a soft grip with the column and provide a stable seal
TRUSTED_SUSTAINABILITY
Specifications
Description 0.25 in. O.D.
For Use With (Equipment) For Varian, Perkin-Elmer