Learn More
10-Phenyl-1-decanol, 96%, Thermo Scientific™ Product Code.: 11328335
10-Phenyl-1-decanol, 96%, Thermo Scientific™
Product Code. 11328335
1 g
Click to view available options
Quantity:
1 g
5 g
Product Code. 11328335
Brand: Thermo Scientific Alfa Aesar L10269.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62607-69-6
Molecular Formula C16H26O
Molecular Weight (g/mol) 234.383
MDL Number MFCD00014426
InChI Key WNFXODBOMMSELY-UHFFFAOYSA-N
Synonym 10-phenyl-1-decanol, benzenedecanol, acmc-20aozo, 10-phenyl-n-decanol, 10-phenyl-1-decyl alcohol
PubChem CID 568845
IUPAC Name 10-phenyldecan-1-ol
SMILES C1=CC=C(C=C1)CCCCCCCCCCO
Specifications
Melting Point 35°C to 37°C
Boiling Point 145°C to 147°C (0.5 mmHg)
Quantity 1 g
Beilstein 1955067
Formula Weight 234.39
Percent Purity 96%
Chemical Name or Material 10-Phenyl-1-decanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC31W003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10142273 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10158537 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberE13U018 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
RUO – Research Use Only
Product Content Correction
Product Title
10-Phenyl-1-decanol, 96%, Thermo Scientific™ >