Learn More
Lenz Laborglasintrumente 1-way Stopcock with PTFE Key Bore
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02162408
Product Code. 10132141
  • € 92.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Description
  • Stopcocks, straight bore, with PTFE plug
  • Acc. to DIN 12541-1
  • With PTFE plug and screwthread retaining device
  • One straight bore
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Specifications
8 mm
Borosilicate Glass (3.3)