missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, ACS, 99+%, Thermo Scientific Chemicals

Filtered through 0.2μ filters. Suitable for spectrophotometry, chromatography

€ 39.10 - € 360.00

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030898.AK
250 mL
€ 39.10
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11337378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030898.K2
1 L
€ 100.00
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11317378
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030898.K7
4 L
€ 360.00
4 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71-41-0
88.15
AMQJEAYHLZJPGS-UHFFFAOYSA-N
6276
pentan-1-ol
C5H12O
MFCD00002977,MFCD00081734,MFCD01075169
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
CHEBI:44884
CCCCCO

Specifications

71-41-0
Colorless
136°C to 138°C
Mild
CH3(CH2)4OH
MFCD00002977,MFCD00081734,MFCD01075169
1730975
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
AMQJEAYHLZJPGS-UHFFFAOYSA-N
pentan-1-ol
6276
88.15
ACS
1-Pentanol
-79°C
0.812 g/mL
49°C (120°F)
C5H12O
1.4103
UN1105
14,7118
Miscible with water,alcohol,ether,acetone and most organic solvents.
CCCCCO
88.15
CHEBI:44884
≥99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

missing translation for 'einecsNumber' : 200-752-1

missing translation for 'rtecsNumber' : SB9800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only