missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 11433797
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate monosodium salt monohydrate acts as a substrate for determination of prostatic acid phosphatase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-89-7
Molecular Formula C10H7O4P
Molecular Weight (g/mol) 222.14
MDL Number MFCD00150615
InChI Key YNXICDMQCQPQEW-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc
PubChem CID 45055387
IUPAC Name naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
SMILES [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 186°C to 191°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 264.16 (246.14 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 1-Naphthyl phosphate monosodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.