missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentene, 98%, Thermo Scientific Chemicals

€ 61.40 - € 769.00

Chemical Identifiers

CAS 693-89-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00001397
InChI Key ATQUFXWBVZUTKO-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
PubChem CID 12746
IUPAC Name 1-methylcyclopentene
SMILES CC1=CCCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423098
View Documents
Thermo Scientific Alfa Aesar
A18816.06
5 g
€ 61.40
5g
Please to purchase this item. Need a web account? Register with us today!
11433098
View Documents
Thermo Scientific Alfa Aesar
A18816.14
25 g
€ 202.00
25g
Please to purchase this item. Need a web account? Register with us today!
11443098
View Documents
Thermo Scientific Alfa Aesar
A18816.22
100 g
€ 769.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

693-89-0
82.15
ATQUFXWBVZUTKO-UHFFFAOYSA-N
12746
CC1=CCCC1
C6H10
MFCD00001397
1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
1-methylcyclopentene

Specifications

693-89-0
0.779
−17°C (1°F)
1.431
UN3295
1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
CC1=CCCC1
82.15
82.15
1-Methylcyclopentene
-142°C
73°C to 75°C
C6H10
MFCD00001397
1900640
ATQUFXWBVZUTKO-UHFFFAOYSA-N
1-methylcyclopentene
12746
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H225-H304

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 211-762-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Methylcyclopentene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : 1-Methylcyclopentene

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only