missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals Product Code.: 11313706

1-Methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Product Code. 11313706
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11313706

Brand: Thermo Scientific Alfa Aesar L14831.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-36-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00144943
InChI Key UQFQONCQIQEYPJ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255Show MoreShow Less
PubChem CID 70255
ChEBI CHEBI:59025
IUPAC Name 1-methylpyrazole
SMILES CN1C=CC=N1

Specifications

Density 0.988
Boiling Point 127°C
Flash Point 35°C (95°F)
Odor Pungent
Refractive Index 1.476
Quantity 5 g
UN Number UN1993
Beilstein 105207
Formula Weight 82.11
Percent Purity ≥97%
Chemical Name or Material 1-Methyl-1H-pyrazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251230 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14831.03,L14831.14,L14831.06
Lot Number10246762 Certificate TypeCertificate of Analysis Date25/05/2023 Catalog NumberL14831.03,L14831.14,L14831.06
Lot Number10173443 Certificate TypeCertificate of Analysis Date08/05/2023 Catalog NumberL14831.03,L14831.14,L14831.06
Lot Number10242363 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberL14831.03,L14831.14,L14831.06
Lot Number10153516 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14831.03,L14831.14,L14831.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.