missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene, 99%, AcroSeal™, Thermo Scientific Chemicals

€ 22.91 - € 122.61

Chemical Identifiers

CAS 592-41-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.15
MDL Number MFCD00009505
InChI Key LIKMAJRDDDTEIG-UHFFFAOYSA-N
Synonym 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
PubChem CID 11597
ChEBI CHEBI:24579
IUPAC Name hex-1-ene
SMILES CCCCC=C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10329437
View Documents
Thermo Scientific Acros
120751000
100 mL AcroSeal™ Glass Bottle
€ 22.91
100mL
Please to purchase this item. Need a web account? Register with us today!
15149163
View Documents
Thermo Scientific Acros
120750010
1 L AcroSeal™ Glass Bottle
€ 122.61
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

592-41-6
84.15
LIKMAJRDDDTEIG-UHFFFAOYSA-N
11597
hex-1-ene
C6H12
MFCD00009505
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
CHEBI:24579
CCCCC=C

Specifications

592-41-6
100.0
Colorless
62.0°C to 65.0°C
Authentic
C6H12
1.3867 to 1.3887
01,215
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
LIKMAJRDDDTEIG-UHFFFAOYSA-N
hex-1-ene
11597
0.34 mm2/s (40°C)
99%
1-Hexene
98.5
-140.0°C
0.6780g/mL
−26°C
98.5% min. (GC)
CH3(CH2)3CH=CH2
MFCD00009505
0.678
Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
CCCCC=C
84.15
CHEBI:24579
84.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 209-753-1

Product Identifier
  • 1-Hexene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only