Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
€ 105.00 - € 1730.00
Specifications
Product Type | Digital Dispenser Cartridge |
---|---|
Autoclavable | Not Autoclavable |
For Use With (Application) | Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting |
Interface | USB |
Plate Type | 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
17961961
![]() |
Thermo Scientific™
LTR0001 |
1-ch 20μL Cartridges for Multidrop Pico 1 |
€ 1560.00
Pack of 30 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
17971961
![]() |
Thermo Scientific™
LTR0002 |
1-ch 200μL Cartridges for Multidrop Pico 1 |
€ 1650.00
Pack of 30 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
17991961
![]() |
Thermo Scientific™
LTR0003 |
4-ch 200 μL Cartridges for Multidrop Pico 8 |
€ 1730.00
Pack of 20 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
16643853
|
Thermo Scientific™
LTR0003 |
4-ch 200 μL Cartridges for Multidrop Pico 8 |
€ 105.00
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
16653853
|
Thermo Scientific™
LTR0004 |
8-ch 20 μL Cartridges for Multidrop Pico 8 |
€ 105.00
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
17981961
![]() |
Thermo Scientific™
LTR0004 |
8-ch 20 μL Cartridges for Multidrop Pico 8 |
€ 1730.00
Pack of 20 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
- Cost Saving through assay miniaturization
- Improved efficiency and sample integrity
- Powerful, flexible, and easy to use software
Specifications
Digital Dispenser Cartridge | |
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting | |
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm | |
11 pL to 10 μL in seconds | |
100/240 V |
Not Autoclavable | |
USB | |
≤ 8% | |
100 to 240V 50/60Hz | |
50/60 Hz |