missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%, Thermo Scientific Chemicals

Product Code. p-102581 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10068721

Brand: Thermo Scientific Acros 112870050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-33-1
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 285°C to 288°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H18Br2
Linear Formula Br(CH2)9Br
Refractive Index 1.4950 to 1.497
Quantity 5 g
Beilstein 01, 166
Specific Gravity 1.4
Molecular Weight (g/mol) 286.04
Formula Weight 286.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 9-Dibromononane, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.