missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%

Product Code. 15488455
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15488455

Brand: Thermo Scientific Alfa Aesar H33424.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1215564-15-0 | C8H4ClF3N4O2S | 312.651 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215564-15-0
Molecular Formula C8H4ClF3N4O2S
Molecular Weight (g/mol) 312.651
MDL Number MFCD11505058
InChI Key SADUDBJMXGGXDE-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl-2-pyrimidinyl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulfonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl-1h-pyrazole-4-sulphonyl chloride, 1-4-trifluoromethyl pyrimidin-2-yl pyrazole-4-sulfonyl chloride, chloro 1-4-trifluoromethyl pyrimidin-2-yl pyrazol-4-yl sulfone
PubChem CID 51063957
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]pyrazole-4-sulfonyl chloride
SMILES C1=CN=C(N=C1C(F)(F)F)N2C=C(C=N2)S(=O)(=O)Cl

Specifications

Melting Point 97°C to 99°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 312.66
Percent Purity 95%
Chemical Name or Material 1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.