missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Pyridyl)ethylamine, 96%

Product Code. 11351979
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11351979

Brand: Thermo Scientific Alfa Aesar H26047.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56129-55-6 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56129-55-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD01691641,MFCD09256803
InChI Key IQVQNBXPYJGNEA-UHFFFAOYNA-N
Synonym 1-pyridin-3-yl-ethylamine, 1-3-pyridyl ethylamine, 1-pyridin-3-yl ethanamine, 3-1-aminoethyl pyridine, 1-3-pyridinyl ethanamine, 1-pyridin-3-yl ethan-1-amine, alpha-methyl-3-pyridinemethylamine, 3-pyridinemethylamine, alpha-methyl, 1-pyridin-3-ylethyl amine, 1-3-pyridyl ethanamine
PubChem CID 2771688
IUPAC Name 1-pyridin-3-ylethanamine
SMILES CC(N)C1=CC=CN=C1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 122.17
Percent Purity 96%
Chemical Name or Material 1-(3-Pyridyl)ethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.