missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)piperazine monohydrochloride monohydrate, 98%, Thermo Scientific™

Product Code. 11484250
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11484250

Brand: Thermo Scientific Alfa Aesar A17570.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1082699-15-7
Molecular Formula C10H16Cl2N2O
Molecular Weight (g/mol) 251.151
MDL Number MFCD00012761
InChI Key RXPHNGRBTQWKEQ-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl piperazine hydrate hydrochloride, 1-2-chlorophenyl piperazine monohydrochloride monohydrate, c10h13cln2.hcl.h2o, 1-2-chlorophenyl piperazine hydrochloride hydrate, 1-2-chlorophenyl piperazine monohydrochloride monohydrate,, 1-2-chlorophenyl piperazinemonohydrochloridemonohydrate
PubChem CID 20235849
IUPAC Name 1-(2-chlorophenyl)piperazine;hydrate;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=C2Cl.O.Cl

Specifications

Melting Point 167°C to 169°C (anhy.)
Quantity 5 g
Beilstein 5652889
Formula Weight 251.16 (233.14 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1-(2-Chlorophenyl)piperazine monohydrochloride monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.