missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody (4D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-92717CL1
This item is not returnable.
View return policy
Description
Nestin Monoclonal antibody specifically detects Nestin in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Nestin | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ21841, nestin | |
| Recombinant construct, amino acids 317-630 of the human Nestin protein expressed in and purified from E. coli. [UniProt P48681] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 4D11 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction