CAS RN 93987-29-2
CAS RN 93987-29-2
IUPAC Name:
2-methyl-1-(2-methyl-5-nitrobenzenesulfonyl)-1H-1,3-benzodiazole
Synonyms:
2-methyl-1-(2-methyl-5-nitrobenzenesulfonyl)-1,3-benzodiazole
Molecular Weight (g/mol):
331.35
Molecular Formula:
C15H13N3O4S
InChi Key:
ILVCPQPMRPHZSG-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC=CC=C2N1S(=O)(=O)C1=C(C)C=CC(=C1)[N+]([O-])=O