accessibility menu, dialog, popup

Test

CAS RN 916256-65-0

H3CNNNBr

CAS RN 916256-65-0

IUPAC Name: 6-bromo-2-methylpyrazolo[1,5-a]pyrimidine
Synonyms: 6-bromo-2-methylpyrazolo[1,5-a]pyrimidine
Molecular Weight (g/mol): 212.05
Molecular Formula: C7H6BrN3
InChi Key: JUWPPXBKRXAJMY-UHFFFAOYSA-N
SMILES: CC1=NN2C=C(Br)C=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results