CAS RN 894783-71-2
CAS RN 894783-71-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BIBF 1202, MedChemExpress
MedChemExpress BIBF 1202 is the carboxylate metabolite of BIBF 1120 which inhibits VEGFR2 kinase with an IC50 of 62 nM.