CAS RN 83907-40-8
CAS RN 83907-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress SPQ
MedChemExpress SPQ is being used to examine and measure membrane chloride transport mechanisms.