CAS RN 808-26-4
CAS RN 808-26-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sancycline, MedChemExpress
MedChemExpress Sancycline is a rare semi-synthetic tetracycline prepared by hydrogenolysis of the chloro and benzylic hydroxy moieties of Declomycin.