CAS RN 804-30-8
CAS RN 804-30-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fursultiamine, MedChemExpress
MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.