CAS RN 69-33-0
CAS RN 69-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tubercidin Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tubercidin Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tubercidin Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Tubercidin, MedChemExpress
MedChemExpress Tubercidin (7-Deazaadenosine) is an antibiotic obtained from Streptomyces tubercidicus.