accessibility menu, dialog, popup

UserName

CAS RN 652-40-4

FOOOF

CAS RN 652-40-4

IUPAC Name: 4,7-difluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4,7-difluoro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 184.1
Molecular Formula: C8H2F2O3
InChi Key: AVLRPSLTCCWJKC-UHFFFAOYSA-N
SMILES: FC1=C2C(=O)OC(=O)C2=C(F)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)