CAS RN 553-21-9
CAS RN 553-21-9
IUPAC Name:
(3aS,11aR)-6,10-dimethyl-3-methylidene-2H,3H,3aH,4H,5H,8H,9H,11aH-cyclodeca[b]furan-2-one
Synonyms:
costunolide costunlide costunolid isocostunolide costus lactone (+)-costunolide
Molecular Weight (g/mol):
232.32
Molecular Formula:
C15H20O2
InChi Key:
HRYLQFBHBWLLLL-AHNJNIBGSA-N
SMILES:
C\C1=C/CC\C(C)=C\[C@H]2OC(=O)C(=C)[C@@H]2CC1
1
–
3
of
3
results
1
(+)-Costunolide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-Costunolide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Costunolide, MedChemExpress
MedChemExpress Costunolide ((+)-Costunolide) is a naturally occurring sesquiterpene lactone, with antioxidative, anti-inflammatory, antiallergic, bone remodeling, neuroprotective, hair growth promoting, anticancer, and antidiabetic properties. Costunolide can induce cell cycle arrest and apoptosis on breast cancer cells.