accessibility menu, dialog, popup

UserName

CAS RN 55-86-7

HClCH3NClCl

CAS RN 55-86-7

IUPAC Name: hydrogen bis(2-chloroethyl)(methyl)amine chloride
Synonyms: mechlorethamine hydrochloride
Molecular Weight (g/mol): 192.51
Molecular Formula: C5H12Cl3N
InChi Key: QZIQJVCYUQZDIR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(CCCl)CCCl
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)