accessibility menu, dialog, popup

UserName

CAS RN 483-14-7

H3CON(S)OH3COCH3OH3C

CAS RN 483-14-7

IUPAC Name: (12bS)-3,4,10,11-tetramethoxy-7,8,12b,13-tetrahydro-5H-6-azatetraphene
Synonyms: tetrahydropalmatine (-)-tetrahydropalmatine (S)-tetrahydropalmatine tetrahydropalmatine (6CI) L-tetrahydropalmatine (S)-(-)-tetrahydropalmatine
Molecular Weight (g/mol): 355.43
Molecular Formula: C21H25NO4
InChi Key: AEQDJSLRWYMAQI-KRWDZBQOSA-N
SMILES: COC1=CC2=C(C=C1OC)[C@@H]1CC3=C(CN1CC2)C(OC)=C(OC)C=C3
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results