CAS RN 477-43-0
CAS RN 477-43-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dehydrocostus Lactone, MedChemExpress
MedChemExpress Dehydrocostus Lactone is a major sesquiterpene lactone isolated from the roots of Saussurea lappa.