accessibility menu, dialog, popup

UserName

CAS RN 372523-75-6

H3CCH3H3CONOCH3OCH3NHNHCl

CAS RN 372523-75-6

IUPAC Name: 1-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)-3,3-dimethyl-2-{[(pyridin-4-yl)methyl]amino}butan-1-one hydrochloride
Synonyms: 1-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-3,3-dimethyl-2-[(pyridin-4-ylmethyl)amino]butan-1-one hydrochloride
Molecular Weight (g/mol): 433.98
Molecular Formula: C23H32ClN3O3
InChi Key: NHKNHFJTMINMBP-UHFFFAOYNA-N
SMILES: Cl.COC1=C(OC)C=C2CN(CCC2=C1)C(=O)C(NCC1=CC=NC=C1)C(C)(C)C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results