accessibility menu, dialog, popup

UserName

CAS RN 36396-99-3

OHO(R)OH(S)OHNNHNNN

CAS RN 36396-99-3

IUPAC Name: (3R,4S)-2-[6-(cyclohexylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (3R,4S)-2-[6-(cyclohexylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 349.39
Molecular Formula: C16H23N5O4
InChi Key: SZBULDQSDUXAPJ-WXURJZIFSA-N
SMILES: OCC1OC([C@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCCC3)N=CN=C12
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results