accessibility menu, dialog, popup

UserName

CAS RN 21746-40-7

CH3NOO

CAS RN 21746-40-7

IUPAC Name: 1-propyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-propylpyrrole-2,5-dione
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: DABFKTHTXOELJF-UHFFFAOYSA-N
SMILES: CCCN1C(=O)C=CC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results