accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103616-89-3

CAS RN 103616-89-3

IUPAC Name: (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: CVZZNRXMDCOHBG-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=CC=C1Cl)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.