accessibility menu, dialog, popup

Test

CAS RN 103476-24-0

CAS RN 103476-24-0

IUPAC Name: hexapotassium 2,3,5-trihydroxy-4,6-bis(phosphonatooxy)cyclohexyl phosphate
Synonyms: hexapotassium 2,3,5-trihydroxy-4,6-bis(phosphonatooxy)cyclohexyl phosphate
Molecular Weight (g/mol): 648.63
Molecular Formula: C6H9K6O15P3
InChi Key: DVOPWCOAGWTTEN-UHFFFAOYNA-H
SMILES: [K+].[K+].[K+].[K+].[K+].[K+].OC1C(O)C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C(O)C1OP([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.