accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1034566-13-6

CAS RN 1034566-13-6

IUPAC Name: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 195.16
Molecular Formula: C6H4F3NOS
InChi Key: FBQFBBGXQZDOQV-UHFFFAOYSA-N
SMILES: CC1=NC(=C(S1)C=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.