CAS RN 10277-43-7
CAS RN 10277-43-7
Lanthanum(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+);trinitrate;hexahydrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Promotions are available
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, REacton™, 99.999% (REO), Thermo Scientific Chemicals
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, Honeywell Fluka™
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+);trinitrate;hexahydrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O