accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102297-41-6

CAS RN 102297-41-6

IUPAC Name: tert-butyl N-[(pyridin-3-yl)methyl]carbamate
Synonyms: tert-butyl N-(pyridin-3-ylmethyl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: PHQXGCNECRISKB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.