accessibility menu, dialog, popup

Test

CAS RN 102-86-3

CH3NCH3CH3

CAS RN 102-86-3

IUPAC Name: trihexylamine
Synonyms: 1-hexanamine, N,N-dihexyl-
Molecular Weight (g/mol): 269.52
Molecular Formula: C18H39N
InChi Key: DIAIBWNEUYXDNL-UHFFFAOYSA-N
SMILES: CCCCCCN(CCCCCC)CCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results