accessibility menu, dialog, popup

Test

CAS RN 1017793-21-3

CAS RN 1017793-21-3

IUPAC Name: 4-bromo-3-chloro-2-fluoropyridine
Synonyms: 4-bromo-3-chloro-2-fluoropyridine
Molecular Weight (g/mol): 210.43
Molecular Formula: C5H2BrClFN
InChi Key: UMQBGWKWOLRROL-UHFFFAOYSA-N
SMILES: FC1=NC=CC(Br)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results