accessibility menu, dialog, popup

Test

CAS RN 101664-59-9

H3CONClOCH3HCl

CAS RN 101664-59-9

IUPAC Name: 2-chloro-3,4-dimethoxypyridine hydrochloride
Synonyms: 2-chloro-3,4-dimethoxypyridine hydrochloride
Molecular Weight (g/mol): 210.05
Molecular Formula: C7H9Cl2NO2
InChi Key: AOBQXXVCWDAJFE-UHFFFAOYSA-N
SMILES: Cl.COC1=C(OC)C(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results