accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1013101-36-4

CAS RN 1013101-36-4

IUPAC Name: 2-amino-8-[4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxypyridin-3-yl)-4-methyl-7H,8H-pyrido[2,3-d]pyrimidin-7-one
Synonyms: 2-amino-8-[4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxypyridin-3-yl)-4-methylpyrido[2,3-d]pyrimidin-7-one
Molecular Weight (g/mol): 425.49
Molecular Formula: C22H27N5O4
InChi Key: XDLYKKIQACFMJG-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=C1)C1=CC2=C(C)N=C(N)N=C2N(C2CCC(CC2)OCCO)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.