CAS RN 10117-38-1
CAS RN 10117-38-1
IUPAC Name:
dipotassium sulfite
Synonyms:
potassium sulfite
Molecular Weight (g/mol):
158.25
Molecular Formula:
K2O3S
InChi Key:
BHZRJJOHZFYXTO-UHFFFAOYSA-L
SMILES:
[K+].[K+].[O-]S([O-])=O
1
–
2
of
2
results
1
2
Potassium sulfite, 90%, pure, Thermo Scientific Chemicals
CAS: 10117-38-1 Molecular Formula: K2O3S Molecular Weight (g/mol): 158.26 InChI Key: BHZRJJOHZFYXTO-UHFFFAOYSA-L Synonym: potassium sulfite,dipotassium sulfite,potassium sulphite,sulfurous acid, dipotassium salt,unii-015kzc652e,sulfurous acid, potassium salt 1:2,potassium sulfite k2so3,kaliumsulfit,potassiumsulfite,dipotassium ion sulfite PubChem CID: 24958 IUPAC Name: dipotassium;sulfite SMILES: [O-]S(=O)[O-].[K+].[K+]
Promotions are available