man

Test

CAS RN 100-70-9

NN

CAS RN 100-70-9

IUPAC Name: pyridine-2-carbonitrile
Synonyms: 2-cyanopyridine 2-pyridyl nitrile picolinic acid nitrile picolinonitrile (8CI) picolinonitrile
Molecular Weight (g/mol): 104.11
Molecular Formula: C6H4N2
InChi Key: FFNVQNRYTPFDDP-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=N1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)