man

Test

CAS RN 100-36-7

H3CNH3CNH2

CAS RN 100-36-7

IUPAC Name: (2-aminoethyl)diethylamine
Synonyms: (2-aminoethyl)diethylamine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: UDGSVBYJWHOHNN-UHFFFAOYSA-N
SMILES: CCN(CC)CCN
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)