man

Test

CAS RN 100-22-1

H3CNCH3NCH3H3C

CAS RN 100-22-1

IUPAC Name: N1,N1,N4,N4-tetramethylbenzene-1,4-diamine
Synonyms: wurster's blue
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: CJAOGUFAAWZWNI-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)